Learn More
eMolecules​ 71989-23-6 | Fmoc-L-isoleucine | Chem-Impex | MFCD00037125 | 353.418 | C21H23NO4 | 0.000 | CC[C@H](C)[C@H](NC(=O)OCC1c2ccccc2-c2ccccc12)C(O)=O | 25g | 112443355
Supplier:  eMolecules​ 112443355
Encompass_Preferred
Fmoc-L-isoleucine | Chem-Impex | 71989-23-6 | MFCD00037125 | 353.418 | C21H23NO4 | 0.000 | CC[C@H](C)[C@H](NC(=O)OCC1c2ccccc2-c2ccccc12)C(O)=O | 25g | 112443355
Catalog No. 50-004-92361