Learn More
Medchemexpress LLC HY-P0175 1mg Medchemexpress, 740 Y-P CAS:1236188-16-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP01751MG
Encompass_Preferred
Medchemexpress, HY-P0175 1mg 740 Y-P CAS:1236188-16-1 740 Y-P (740YPDGFR; PDGFR 740Y-P) is a potent and cell-permeable PI3K activator. 740 Y-P readily binds GST fusion proteins containing both the N- and C- terminal SH2 domains of p85 but fails to bind GST alone. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9430