Learn More
Selleck Chemical LLC 740 Y-P
SDP
Supplier:  Selleck Chemical LLC S7865100MG
Encompass_Preferred
740 Y-P (PDGFR 740Y-P 740YPDGFR) is a cell-permeable phosphopeptide activator of PI3K
Catalog No. 50-000-00574