Learn More
Selleck Chemical LLC 740 Y-P (PDGFR 740Y-P) 1mg 1236188-16-1
SDP
Supplier:  Selleck Chemical LLC S78651MG
Encompass_Preferred
740 Y-P is cell-permeable phosphopeptide activator of PI3K. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5355