Learn More
COANN TECHNOLOGIES LLC High-quality 50-75 um inner diameter nanoLC column
Supplier:  COANN TECHNOLOGIES LLC HEB07503001718IWF
Encompass
High-quality 50-75 um inner diameter nanoLC columns for conventional mass spectrometry of peptides (proteomics) and metabolites with fittings | 75 um ID x 30 cm L x 365 um OD | 1.7 um C18 (100A) | 200 ± 50 nL/min | <5000 psi | With Fittings | Integrated
Catalog No. NC1599906