Learn More
Nanocomposix NanoXact Silver Nanocubes - PVP - 75 nm, 1 mg/mL in ethanol, 5 mL
SDP
Supplier:  Nanocomposix SCPH755M
Encompass_Preferred
As a global leader in silver nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.75 nm silver nanocubes combine distinctive anisotropic morphology and unique plasmonic optical properties. These nanoparticles are useful in a broad range of applications from biosensors, sensing, imaging, SERS, metamaterials, and more.nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All products are supplied with batch-specific CoA's including TEM, DLS, Zeta, and UV-Vis characterization data.Intended for Research Use Only. Contact us for custom formulations, ISO 13485/cGMP compliant materials, or bulk supply requirements.
Catalog No. NC2885971