Learn More
Ace Glass, Inc. 7621 CAP W/HOLE GL14 7621 CAP W/HOLE GL14
SDP
Supplier:  Ace Glass, Inc. 762104
Encompass
GL14 Duran, polybutylene terephthalate (PBT) open-top caps with center hole. PBT temperature range is -45 - 180¿C. Can be used with Duran hose connectors, Duran sealing ring for GL thread hose connectors, and silicone or silicone/PTFE septa for Duran caps.
Catalog No. NC9957796