Learn More
Cayman Chemical 7Ethoxy4 trIfluoromethylc 50mg
SDP
Supplier:  Cayman Chemical 3461050 MG
Encompass_Preferred
A fluorogenic substrate for CYPs; HFC is released upon O-deethylation by CYPs and its fluorescence can be used to quantify CYP activity; ex/em = 410/510 nm, respectively
Catalog No. 50-294-6886