Learn More
Medchemexpress LLC 7H-dibenzo[c,g]carbazole | 194-59-2 | 100.0% | 25 G
SDP
Supplier:  Medchemexpress LLC HY11998325G
Encompass_Preferred
7H-Dibenzo[c,g]carbazole is an azaarene research chemical used in toxicology and metabolic activation studies. It is a white to off-white solid with molecular formula C20H13N and molecular weight 267.32 g/mol. Reported high purity makes it suitable for analytical and toxicological experiments.
  • High purity suitable for analytical and toxicological studies.
  • Solid, white to off-white appearance for easy handling.
  • Used to study DNA adduct formation and metabolic activation.
  • Available in multiple pack sizes for flexible ordering.
  • Molecular formula C20H13N and molecular weight 267.32 g/mol.
Catalog No. 50-002-32442