Learn More
Medchemexpress LLC 7H-Pyrrolo[2,3-d]pyrimidin-4-amine, N-[2-(1-azetidinylsulfonyl)phenyl]-7-methyl-2-(3-methyl | 3055550-22-3 | 99.9% | 423.49 g·mol⁻¹ | C20H21N7O2S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14953010MG
Encompass_Preferred
EGFR-IN-86 is a small-molecule epidermal growth factor receptor (EGFR) inhibitor reported to have an IC50 of 1.5 nM. It has demonstrated activity in glioblastoma models, inducing apoptosis and causing G2/M cell-cycle arrest in U87 cells. Supplied as a high-purity research compound (99.92%), it is intended for in vitro and preclinical studies. Molecular formula C20H21N7O2S; molecular weight 423.49 g·mol⁻¹.
  • Potent EGFR inhibition with a reported IC50 of 1.5 nM.
  • Demonstrated activity in glioblastoma models, inducing apoptosis.
  • Induces G2/M cell-cycle arrest in U87 cells.
  • High purity suitable for research applications.
  • Supplied in small research-friendly quantities for laboratory use.
  • Molecular formula C20H21N7O2S and molecular weight 423.49 g·mol⁻¹.
Catalog No. 50-002-85839