Learn More
Cayman Chemical 7Hydroxy4methyl8nItrocou 500mg
SDP
Supplier:  Cayman Chemical 30900500 MG
Encompass_Preferred
A coumarin derivative with antibacterial and antioxidant activities; active against S. aureus and E. coli in a disc assay at 50 and 100 UG/disc; inhibits NADPH-dependent lipid peroxidation in rat liver microsomes
Catalog No. 50-298-9880