Learn More
Chemglass Life Sciences Chemglass 7 L Bio Reactor Baffle Assembly, M10 (CLS-1383-11)
SDP
Supplier:  Chemglass Life Sciences CLS138311
Encompass
Replacement baffle assembly designed for 7 L bioreactors. Baffles mount near the reactor wall to disrupt vortex formation and improve mixing efficiency during fermentation and cell-culture processes. The assembly fastens using an M10 thread and is intended as a direct replacement in standard 7 L reactor systems.
  • Baffle purpose: increase mixing efficiency inside the reactor.
  • Compatible volume: 7 L bioreactors.
  • Thread size: M10.
  • Mounting location: near reactor wall for optimal mixing performance.
  • Maintenance: replacement silicone o-ring available for seal maintenance.
Catalog No. 50-273-3313