Learn More
Cayman Chemical 7Methoxy4 trIfluoromethyl
SDP
Supplier:  Cayman Chemical 34398250 MG
Encompass_Preferred
A fluorogenic substrate for CYPs; HFC is released upon enzymatic cleavage by CYPs and its fluorescence can be used to quantify CYP activity; ex/em = 410/510 nm, respectively
Catalog No. 50-294-6172