Learn More
Leica Biosystems 7ml SMA Bond RTU Primary
Supplier:  Leica Biosystems PA0943U
Encompass
Antigen BackgroundCytoplasmic actins are part of the microfilament system of cytoskeletal proteins. Smooth muscle actin is found in vascular walls, intestinal muscularis mucosae and muscularis propria and in the stroma of various tissues.Product Specific InformationEnzyme pretreatment may enhance staining in some cases.DisclaimerSMA (Alpha Smooth Muscle Actin) is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2016