Learn More
Ace Glass, Inc. #7 Ace-Thred solid nylon plug, 7855-707 FETFE O-ring front seal
SDP
Supplier:  Ace Glass, Inc. 584604
Encompass_Preferred
#7 Ace-Thred solid nylon plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -009 FETFE O-ring.
Catalog No. 50-955-7159