Learn More
Medchemexpress LLC (7S)-3-fluoro-4-[3-(8-fluoro-1-methyl-2,4-dioxoquinazolin-3-yl)-2-methylphenyl]-7-(2-hydroxypropan... | 1643368-58-4 | 99.4% | 572.60 g·mol⁻¹ | C32H30F2N4O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10185625MG
Encompass_Preferred
BMS-986142 is a potent, highly selective, reversible small-molecule inhibitor of Bruton's tyrosine kinase (BTK) with reported IC50 ≈ 0.5 nM. It is supplied as a high-purity research reagent (powder) for preclinical and translational studies; molecular formula C32H30F2N4O4 and CAS 1643368-58-4.
  • Potent reversible BTK inhibition (IC50 ≈ 0.5 nM).
  • High selectivity across kinase panels to reduce off-target effects.
  • High purity suitable for biochemical and cellular assays.
  • Stable powder form with defined storage conditions for long-term use.
  • Suitable for preclinical pharmacology and mechanistic studies.
Catalog No. 50-002-48081