Learn More
Sterlitech Corporation Polyester (PETE) Membrane Filters, 8.0 Micron, 25mm, 100/Pk
SDP
Supplier:  Sterlitech Corporation PET8025100
Encompass_Preferred
Sterlitech polyester track etch (PETE) membrane filters, 8 micron pore size, 25 mm diameter, pack of 100. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.
Catalog No. NC1750599