Learn More
Cayman Chemical 8 12-Iso-IPF2a-VI 25ug
SDP
Supplier:  Cayman Chemical 1631025 UG
Encompass_Preferred
The most abundant isoprostane product formed during lipid peroxidation; detected after CCl4-induced oxidative damage to hepatic tissue and has been found elevated in the urine, blood, and CSF of patients with Alzheimer’s disease
Catalog No. 50-297-2636