Learn More
Medchemexpress LLC 8-[(1R)-1-(3,5-difluoroanilino)ethyl]-N,N-dimethyl-2-morpholinyl-4-oxo-chromene-6-carboxamide | 1627494-13-6 | MFCD27987908 | 100.0% | 457.5 g/mol | C24H25F2N3O4 | 50MG
SDP
Supplier:  Medchemexpress LLC HY1233050MG
Encompass_Preferred
AZD8186 is a selective, small-molecule inhibitor of phosphoinositide 3-kinase beta (PI3Kβ) and delta (PI3Kδ) used as a research compound to probe PI3K signaling pathways in oncology research. It exhibits low-nanomolar potency against PI3Kβ and PI3Kδ, selectivity over other PI3K isoforms, and has been evaluated in preclinical models and early clinical studies.
  • Low-nanomolar potency against PI3Kβ and PI3Kδ (IC50 ≈ 4 nM and 12 nM).
  • Selectivity over PI3Kα and PI3Kγ to support targeted pathway studies.
  • High stated purity suitable for research applications.
  • Available in multiple small-scale package sizes for laboratory use.
  • Well-characterized chemical identifiers for database cross-referencing.
Catalog No. 50-002-37461