Learn More
Medchemexpress LLC 8-(6-Methoxypyridin-3-yl)-1-(4-(piperazin-1-yl)-3-(trifluoromethyl)phenyl)-1,5-dihydro-4H-triazolo | 2648986-65-4 | 99.9% | 521.49 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14765550MG
Encompass_Preferred
CQ211 is a potent and selective RIOK2 inhibitor with a Kd of 6.1 nM. It exhibits potent proliferation inhibition activity against multiple cancer cell lines. In vitro, CQ211 suppresses the phosphorylation of mTOR in cancer cells. It effectively inhibits the proliferation of MKN-1 and HT-29 cells, with IC50 values of 0.61 μM and 0.38 μM, respectively. In vivo, administration of CQ211 at 25 mg/kg was observed to inhibit the MKN-1 xenograft mouse model, resulting in a tumor growth inhibition (TGI) of 30.9%.
  • Potent and selective RIOK2 inhibitor
  • Exhibits proliferation inhibition activity against multiple cancer cell lines
  • Suppresses phosphorylation of mTOR in cancer cells
  • Inhibits MKN-1 and HT-29 cell proliferation
  • Demonstrates tumor growth inhibition in xenograft mouse models
Catalog No. 50-003-61545