Learn More
Cayman Chemical 8 9-EE-14 Z-E 100ug
SDP
Supplier:  Cayman Chemical 18112100 UG
Encompass_Preferred
An oxylipin and a metabolite of arachidonic acid; increases glomerular membrane albumin permeability and reduces the membrane protective effects of 8(9)-EET in isolated rat glomeruli at 300 nM
Catalog No. 50-296-8394