Learn More
Cayman Chemical 8 9DiHETE Fatty Acids and Deri
SDP
Supplier:  Cayman Chemical 1047350 UG
Encompass_Preferred
A major metabolite of EPA; produced in liver microsomes by CYP450 monooxygenases
Catalog No. 50-288-9689