Learn More
Cayman Chemical 8 9epoxy HexahydrocnabIn 100ug
SDP
Supplier:  Cayman Chemical 42022100 UG
Encompass_Preferred
An analytical reference standard categorized as a phytocannabinoid metabolite; an active metabolite of Δ8-THC; prolongs pentobarbital-induced sleeping time in mice; intended for research and forensic applications
Catalog No. 50-294-3115