Learn More
AdipoGen 8-Acetoxypyrene-1 3 6-trisulfo
SDP
Supplier:  AdipoGen CDXA0172M200
Encompass_Preferred
Chemical. CAS 115787-83-2. Formula C18H9Na3O11S3. MW 566.42. Synthetic. Highly water soluble enzyme substrate for the fluorometric assay of esterases at longer wavelengths.
Catalog No. 50-256-1819