Learn More
Abcam 8-Aceto x ypyrene-1,3,6-trisulfonic acid trisodium salt, Fluorogenic esterase substrate, 200MG
Supplier:  Abcam AB145400200MG
Encompass_Preferred
MW 566.4 Da, Purity >98%. Highly water soluble substrate for fluorometric assay of esterases at longer wavelengths.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5228