Learn More
Medchemexpress LLC 8-Aminoadenine | 28128-33-8 | 98.3% | 150.14 g·mol⁻1 | C5H6N6 | 10 MG

Supplier: Medchemexpress LLC HY10317210MG
8-Aminoadenine is a small-molecule adenine receptor ligand used in biochemical and pharmacological research. The compound is reported to have a Ki of 6.51 μM at the human binding site and is supplied as a solid for assay development, binding studies, and small-scale synthesis.
- Chemical formula C5H6N6 and molecular weight 150.14 g·mol⁻1.
- High purity (98.34%) suitable for research applications.
- Available in small research pack sizes (example: 10 MG) for screening and method development.
- Solid appearance, brown to reddish brown, with specified storage conditions for stability.
- Analytical documentation available: datasheet, COA, HNMR, LCMS, and RP-HPLC.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.