Learn More
Medchemexpress LLC 8-aminoadenosine | 3868-33-5 | MFCD00047230 | 99.9% | 282.26 | C10H14N6O4 | 50MG

Supplier: Medchemexpress LLC HY12592750MG
8-Aminoadenosine is an RNA-directed nucleoside analogue used in cellular and biochemical research. It reduces cellular ATP levels, inhibits mRNA synthesis, and interferes with signaling pathways to induce autophagy and apoptosis, making it useful for studies of transcriptional inhibition, energy metabolism, and anticancer mechanisms.
- Reduces cellular ATP levels and inhibits mRNA synthesis.
- Blocks Akt/mTOR signaling and induces autophagy and apoptosis.
- High purity (≈99.9%).
- Soluble in DMSO (≈83.3 mg/mL); may require sonication.
- Available in small research sizes (5-200 mg).
- Storage: powder -20°C (long term); in solution -80°C (short term).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.