Learn More
Medchemexpress LLC 8-aminoadenosine | 3868-33-5 | MFCD00047230 | 99.9% | 282.26 g/mol | C10H14N6O4 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12592725MG
Encompass_Preferred
8-Aminoadenosine is an RNA-directed adenosine nucleoside analogue used in biochemical and pharmacological research to reduce cellular ATP levels and inhibit mRNA synthesis. It is supplied as a white to off-white solid with a reported molecular formula of C10H14N6O4 and molecular weight of 282.26 g/mol. The compound is soluble in DMSO and requires appropriate cold storage for long-term stability.
  • Reduces cellular ATP and inhibits mRNA synthesis.
  • Applicable for studies of RNA metabolism and cell signaling.
  • High reported purity (99.94%).
  • White to off-white solid, suitable for powder handling.
  • Soluble in DMSO; sonication may improve solubility.
  • Stable when stored under recommended freezer conditions.
Catalog No. 50-002-36514