Learn More
Medchemexpress LLC 8-Aminopyrene-1,3,6-trisulfonic acid trisodium (APTS) | 196504-57-1 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYD0031100MG
Encompass_Preferred
8-Aminopyrene-1,3,6-trisulfonic acid trisodium (APTS) is an anionic fluorescent dye with excitation/emission at 425/503 nm in pH 7.4 PBS. Its fluorescence intensity remains stable between pH 4-10. This compound retains its fluorescent properties when immobilized in a hydrogel through copolymerization and can be utilized for glucose sensing.
  • Anionic fluorescent dye
  • Excitation/emission at 425/503 nm in pH 7.4 PBS
  • Fluorescence intensity stable between pH 4-10
  • Retains fluorescent properties when immobilized in a hydrogel through copolymerization
  • Utilized for glucose sensing
  • Exhibits an apparent glucose binding constant of 30 M-1 at an optimal Q:D ratio (13:1)
  • Increases signal by 14% upon increasing glucose concentration from 0 to 30 mM
Catalog No. 50-004-10791