Learn More
Medchemexpress LLC 8-bromo-cAMP sodium salt | 76939-46-3 | MFCD00005844 | 99.9% | 430.08 | C10H10BrN5NaO6P | 25MG
SDP
Supplier:  Medchemexpress LLC HY1230625MG
Encompass_Preferred
8-Bromo-cAMP sodium salt is a membrane-permeable analog of cyclic adenosine monophosphate (cAMP) that activates cyclic AMP-dependent protein kinase (PKA). It is used in biochemical and cell biology research to mimic cAMP signaling and probe PKA-mediated pathways involved in regulation of proliferation, differentiation, and apoptosis. Supplied as a sodium salt powder with high purity suitable for in vitro assays.
  • Cell-permeable cAMP analog for intracellular signaling studies.
  • Activates cyclic AMP-dependent protein kinase (PKA).
  • High purity suitable for biochemical and cellular assays.
  • Stable sodium salt form for convenient handling and solution preparation.
  • Available in small quantities for lab-scale experiments (e.g., 25 mg).
Catalog No. 50-002-38386