Learn More
Medchemexpress LLC 8-bromo-cGMP (sodium) | 51116-01-9 | 99.9% | 446.08 | C10H10BrN5NaO7P | 1 ML
SDP
Supplier:  Medchemexpress LLC HY101379A10MM 1ML
Encompass_Preferred
8-Bromo-cGMP sodium is a membrane-permeable analogue of cyclic guanosine monophosphate (cGMP) used to activate protein kinase G (PKG) in biochemical and cellular assays. It is available as a ready-to-use 10 mM solution (1 mL) and in solid quantities for custom preparation.
  • Membrane-permeable analogue of cGMP for PKG activation.
  • Ready-to-use 10 mM solution format simplifies dosing.
  • Also available as solid quantities for custom preparation.
  • High purity (~99.9%) for reliable experimental results.
  • Well characterized with molecular weight 446.08 and CAS 51116-01-9 for identification.
Catalog No. 50-002-13202