Learn More
Medchemexpress LLC 8-bromo-cGMP sodium | 51116-01-9 | 99.9% | 446.08 | C10H10BrN5NaO7P | 25 MG
SDP
Supplier:  Medchemexpress LLC HY101379A25MG
Encompass_Preferred
8-Bromo-cGMP sodium is a membrane-permeable analogue of cyclic guanosine monophosphate (cGMP) that acts as a protein kinase G (PKG) activator. It is used in biochemical and cellular research to probe cGMP-dependent signaling pathways involved in vascular tone, nociception, calcium channel modulation, and insulin secretion.
  • Membrane-permeable cGMP analogue for intracellular delivery
  • Activates protein kinase G (PKG) in functional assays
  • Modulates calcium channel activity and insulin secretion pathways
  • Useful for studies of vasodilation and nociception
  • Available as solid and aqueous solution formats
  • High purity suitable for research applications
Catalog No. 50-002-13230