Learn More
Medchemexpress LLC 8-Bromo-cGMP sodium | 51116-01-9 | 99.9% | 446.08 g/mol | C10H10BrN5NaO7P | 50 MG
SDP
Supplier:  Medchemexpress LLC HY101379A50MG
Encompass_Preferred
8-Bromo-cGMP sodium is a membrane-permeable cyclic GMP analogue used to activate protein kinase G (PKG) in biochemical and cellular research. It mimics intracellular cGMP signaling and is commonly used to probe cGMP-dependent pathways in pharmacological and physiological studies.
  • Membrane-permeable cyclic GMP analogue
  • Activates protein kinase G (PKG) in biochemical assays
  • High purity suitable for research use (99.89% per manufacturer)
  • Solid form, off-white to light yellow appearance
  • Available in small solid packages for experimental use
  • Stable when stored sealed and protected from moisture; in solvent: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-002-11809