Learn More
Medchemexpress LLC 8-bromoadenosine 3',5'-cyclic monophosphate | 23583-48-4 | 99.5% | 408.10 | C10H11BrN5O6P | 50MG
SDP
Supplier:  Medchemexpress LLC HY12306A50MG
Encompass_Preferred
8-Bromo-cAMP is a membrane-permeable cyclic AMP analog used to activate cAMP-dependent protein kinase (PKA) and study cAMP/PKA signaling in cellular systems. It is more resistant to phosphodiesterase-mediated degradation than native cAMP, producing prolonged intracellular responses; studies report anti-proliferative and pro-apoptotic effects in some cancer cell models. The compound is supplied as a solid powder and is intended for laboratory research applications.
Catalog No. 50-002-36340