Learn More
Medchemexpress LLC 8-bromoadenosine 3',5'-cyclic monophosphate | 23583-48-4 | MFCD00075580 | 99.5% | 408.10 g/mol | C10H11BrN5O6P | 25MG
SDP
Supplier:  Medchemexpress LLC HY12306A25MG
Encompass_Preferred
8-Bromo-cAMP (8-Br-cAMP) is a membrane-permeable cyclic AMP analogue that activates cyclic AMP-dependent protein kinase (PKA). It is used in biochemical and cell biology research to probe cAMP/PKA signaling pathways and has reported anti-proliferative and pro-apoptotic effects in some cancer cell models. Supplied as a solid, it is commonly used in cell signaling studies, biochemical assays, and mechanistic investigations.
  • Membrane-permeable cAMP analogue.
  • Activates cyclic AMP-dependent protein kinase (PKA).
  • Useful for cell signaling and biochemical assays.
  • Reported anti-proliferative and pro-apoptotic activity in cell studies.
  • High purity suitable for research applications.
  • Available in multiple pack sizes for experimental flexibility.
Catalog No. 50-002-37936