Learn More
Medchemexpress LLC 8-Bromoguanosine | 4016-63-1 | MFCD00037985 | 100.0% | 362.14 g·mol⁻1 | C10H12BrN5O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01903310MM/1ML
Encompass_Preferred
8-Bromoguanosine is a brominated derivative of the purine nucleoside guanosine used in biochemical and structural studies to modulate nucleic acid conformation. Incorporation or interaction of the brominated base can reduce RNA conformational heterogeneity and facilitate structural analysis. The compound is supplied as a high-purity research reagent in solution form for convenient use in assays.
  • Brominated purine nucleoside for biochemical and structural studies.
  • Reduces conformational heterogeneity of RNA to aid structural analysis.
  • Provided as a 10 mM solution in DMSO for convenient assay use.
  • High purity suitable for research applications.
  • Store protected from light and keep frozen to preserve stability.
Catalog No. 50-003-00838