Learn More
Capitol Scientific Inc 8-CHAN X 7ML REAG RES,100/CS
SDP
Supplier:  Capitol Scientific Inc COR4878
Encompass
Corning® 4878 Costar® Transtar-96® 8-Channel x 7mL Reagent Reservoir for Multi-Channel Pipettors, White Polystyrene, Sterile (Case of 100)
Catalog No. NC2200021