Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (8-(Cyclohexyloxy)-1-oxo-2-phenyl-1H-benzo[4,5]thiazolo[3,2-a]pyridine-4-carbonyl)-L-tyrosine | 2245195-67-7 | 96.7% | 582.67 | C33H30N2O6S | 50 MG
SDP

Supplier:  Medchemexpress LLC HY10074950MG

Encompass_Preferred

HeE1-2Tyr is a pyridobenzothiazole small molecule that inhibits RNA-dependent RNA polymerases (RdRp), showing in vitro activity against flavivirus RdRps and SARS-CoV-2.

  • Potent RdRp inhibitor with reported IC50 ≈ 27.6 μM in vitro.
  • Molecular formula C33H30N2O6S and molecular weight 582.67.
  • High purity (96.7%) and solid, white to yellow appearance.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Storage guidance: powder long term at -20 °C; in solvent store at -80 °C for extended stability.
  • Available in small-scale solid quantities, including 50 mg sizes for research use.

Catalog No. 50-002-12863


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.