Learn More
Medchemexpress LLC (8-(Cyclohexyloxy)-1-oxo-2-phenyl-1H-benzothiazolo[3,2-a]pyridine-4-carbonyl)-L-tyrosine | 2245195-67-7 | 96.7% | 582.67 g·mol-1 | C33H30N2O6S | 5 MG

Supplier: Medchemexpress LLC HY1007495MG
HeE1-2Tyr is a pyridobenzothiazole small-molecule inhibitor of flavivirus RNA-dependent RNA polymerase (RdRp) intended for in vitro antiviral research; it is supplied with batch-specific COA and recommended cold storage.
- Inhibits flavivirus RdRp in vitro.
- Demonstrated activity against West Nile and dengue virus.
- Supplied as five mg solid and as 10 mM solution in DMSO.
- Purity typically 96.7% per batch certificate of analysis.
- Recommended storage: powder at -20°C, solutions at -80°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.