Learn More
Abcam 8-Cyclopentyl-1,3-dimethyl x anthine, A1 adenosine receptor antagonist, 1G
Supplier:  Abcam AB1447451G
Encompass_Preferred
MW 248.28 g/mol. Potent, selective A1 adenosine receptor antagonist: binding activity Ki = 10.9 nM (A1 receptor); Ki = 1440 nM (A2 receptor).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5510