Learn More
Medchemexpress LLC 8-Geranyloxypsoralen | 7437-55-0 | 1 MG

Supplier: Medchemexpress LLC HYN22621MG
8-Geranyloxypsoralen is a furanocoumarin isolated from grapefruit. It acts as a potent inhibitor of P450 3A4 (CYP3A4) with an IC50 of 3.93 μM. This product is for research use only and not intended for human use or for sale to patients.
- Solid form, off-white to light yellow
- Purity: 99.93%
- Molecular formula: C21H22O4
- Molecular weight: 338.40
- Store at 4°C, protect from light
- In solvent, store at -80°C for 6 months or -20°C for 1 month, protect from light
- Classified as phenylpropanoid and coumarin
- Initially sourced from plants, Leguminosae, Psoralea corylifolia L.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.