Learn More
Cayman Chemical 8-Gernyloxypsoralen 10mg
SDP
Supplier:  Cayman Chemical 3516710 MG
Encompass_Preferred
A prenylated furanocoumarin with diverse biological activities; inhibits CYP3A4 and BACE1 (IC50s = 3.93 and 20.4 µM, respectively); inhibits TPA-induced Epstein-Barr virus activation in infected Raji cells, TPA-induced superoxide generation in HL-60 cells, and LPS- and IFN-γ-induced NO generation in RAW 264.7 macrophages at 50 µM
Catalog No. 50-291-1169