Learn More
Medchemexpress LLC 8-hydroxy-5-nitroquinoline-d4 | 4008-48-4 | MFCD00006791 | 194.18 | C9H2D4N2O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB1159S10MG
Encompass_Preferred
Nitroxoline-d4 (8-hydroxy-5-nitroquinoline-d4) is the deuterated isotopologue of nitroxoline provided as a reference standard for research and analytical applications. It is used as an internal standard in mass spectrometry and in studies of nitroxoline's antibiofilm and metal-chelating activity.
  • Deuterated internal standard for analytical assays
  • Suitable for mass spectrometry quantitation and method development
  • Supports antibiofilm and antimicrobial research applications
  • High isotopic labeling minimizes overlap with native analyte
  • Molecular formula C9H2D4N2O3, molecular weight 194.18
Catalog No. 50-002-97789