Learn More
Cayman Chemical 8-HydroxygunosIn 10mg
SDP
Supplier:  Cayman Chemical 8930010 MG
Encompass_Preferred
An oxidized nucleoside and a marker of RNA oxidative damage; levels of 8-hydroxyguanosine increase in HeLa cells when cultured with hydrogen peroxide; has been detected in neurons in postmortem brain from patients with Alzheimer's or Parkinson's disease
Catalog No. 50-289-6229