Learn More
Medchemexpress LLC 8-Hydroxyquinoline (8-Quinolinol) | 148-24-3 | >99.9% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB10055G
Encompass_Preferred
8-Hydroxyquinoline (8-Quinolinol) is a lipophilic metal chelator that functions as a fungicide. It exhibits a MIC range of 27.56-55.11 μM (4-8 μg/mL) against clinical isolates of *Neisseria gonorrhoeae*. This compound can bind to copper, forming complexes that facilitate copper transport into cells. Additionally, 8-Hydroxyquinoline leads to an increase in micronucleated polychromatic erythrocytes and can cause hair depigmentation in mice.
  • Functions as a fungicide
  • Exhibits a MIC range of 27.56-55.11 μM (4-8 μg/mL) against clinical isolates of Neisseria gonorrhoeae
  • Binds to copper, forming complexes that facilitate copper transport into cells
  • Can lead to an increase in micronucleated polychromatic erythrocytes
  • May cause hair depigmentation in mice
Catalog No. 50-004-07473