Learn More
Selleck Chemical LLC 8-Hydroxyquinoline S4547-100mg
SDP
Supplier:  Selleck Chemical LLC S4547100MG
Encompass_Preferred
8-Hydroxyquinoline (8-Oxychinolin 8-Quinolinol Oxine) is an antiseptic with mild fungistatic bacteriostatic anthelmintic and amebicidal action It is also used as a reagent and metal chelator as a carrier for radio-indium for diagnostic purposes
Catalog No. 50-311-1491