Learn More
Medchemexpress LLC 8-hydroxyquinoline hemisulfate (8-quinolinol hemisulfate) | 134-31-6 | 100.0% | 194.20 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW012037500MG
Encompass_Preferred
8-Hydroxyquinoline hemisulfate (8-Quinolinol hemisulfate) is a monoprotic bidentate chelating agent that exhibits antiseptic, disinfectant, and pesticide properties, functioning as a transcription inhibitor.
Catalog No. 50-004-53618