Learn More
Medchemexpress LLC 8-Hydroxyquinoline hemisulfate (Standard) | 134-31-6 | 99.6% | 194.20 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW012037R
Encompass_Preferred
8-Hydroxyquinoline hemisulfate (Standard) is an analytical standard and monoprotic bidentate chelating agent. It is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Functions as a transcription inhibitor
  • Exhibits antiseptic properties
  • Displays disinfectant properties
  • Acts as a pesticide
Catalog No. 50-004-52706