Learn More
MSC Thomas & Betts TY5272MX 8-3/4" Long Black Nylon Standard Cable Tie 120 Lb Tensile Strength, 1.27mm Thick, 50.8mm Max Bundle Diam
Supplier:  MSC 54066089
Encompass
Thomas & Betts TY5272MX 8-3/4" Long Black Nylon Standard Cable Tie 120 Lb Tensile Strength, 1.27mm Thick, 50.8mm Max Bundle Diam
Catalog No. 50-129-9827