Learn More
Apexbio Technology LLC 8-Methoxypsoralen 298-81-7 1g
SDP
Supplier:  Apexbio Technology LLC N13051000
Encompass_Preferred
8-Methoxypsoralen (CAS 298-81-7) is a tricyclic furocoumarin compound recognized for its function as a suicide inhibitor of cytochrome P450 (CYP) enzymes By irreversibly binding to CYP active sites 8-Methoxypsoralen effectively impedes enzymatic activity thereby modulating metabolic pathways involving CYPs Comparative studies indicate that 8-Methoxypsoralen demonstrates greater inhibitory potency against CYP enzymes than related furanocoumarins such as 5-MOP 5-hydroxypsoralen dihydro-8-MOP and psoralen This compound is widely employed in research to investigate CYP-mediated metabolism drug-drug interactions and enzyme inhibition mechanisms
Catalog No. 50-224-0308